Tisk…

D-(-)-α-Phenylglycine 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: D-(-)-2-Phenylglycine, H-D-Phg-OH, (R)-(-)-α-Aminophenylacetic acid

130630250 130631000
ACRO130630250EA 1430 CZK
ACRO130630250 ACRO130631000
D-(-)-α-Phenylglycine 98%
D-(-)-α-Phenylglycine
Vzorec: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00008061
CAS číslo: 875-74-1
EINECS: 212-876-3

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Specific optical rotation -151° to -161° (20°C, 589 nm) (c=5, 1M HCl)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR