Tisk…

Girard's reagent T (carbazoylmethyltrimethylammonium chloride) 99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride

119870500 119872500
ACRO119870500EA 1260 CZK
ACRO119870500 ACRO119872500
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) 99%
Girard's reagent T (carbazoylmethyltrimethylammonium chloride)
Vzorec: C₅H₁₄ClN₃O
MW: 167.64
Bod tání: 188…192 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012009
CAS číslo: 123-46-6
EINECS: 204-629-3

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 187 to 192 °C
Titration Argentometric ≥98.5 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR