Tisk…

Diphenyl chlorophosphate 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Diphenyl phosphorochloridate

117240250 117241000 117245000
ACRO117240250EA 1090 CZK
ACRO117240250 ACRO117241000 ACRO117245000
Diphenyl chlorophosphate 98%
Diphenyl chlorophosphate
Vzorec: C₁₂H₁₀ClO₃P
MW: 268,64 g/mol
Bod varu: 314…316 °C (272 mmHg)
Hustota: 1,29 g/cm³
Bod vzplanutí: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003030
CAS číslo: 2524-64-3
EINECS: 219-759-6
UN: 3265
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5485 to 1.5505 (20°C, 589 nm)
GC ≥97.5 %
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meq/g
Thiophene Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR