1-(2-Chloroethyl)piperidinium chloride 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
109211000
109215000
ACRO109211000EA
1320
CZK
ACRO109211000
ACRO109215000
1-(2-Chloroethyl)piperidinium chloride 98%
1-(2-Chloroethyl)piperidinium chloride
Vzorec:
C₇H₁₅Cl₂N Bod tání: 228…234 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012837 CAS číslo: 2008-75-5 EINECS: 217-920-5 UN: 2928 ADR: 6.1,II |
Výsledky specifikačního testu
Appearance | White to yellow to beige Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 228 to 234 °C |
Loss on drying | ≤1 % (105 °C, 3 hrs) |
Additional info | total chloride: 38.0 to 39.0 % w/w |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...