Aminoacetaldehyde diethyl acetal 99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal
103070050
103070500
103072500
ACRO103070050EA
655
CZK
ACRO103070050
ACRO103070500
ACRO103072500
Aminoacetaldehyde diethyl acetal 99%
Aminoacetaldehyde diethyl acetal
Vzorec:
C₆H₁₅NO₂ MW: 133,19 g/mol Bod varu: 162…163 °C (760 mmHg) Hustota: 0,916 g/cm³ Bod vzplanutí: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008136 CAS číslo: 645-36-3 EINECS: 211-439-4 UN: 2733 ADR: 3,III |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4160 to 1.4180 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...