Peroxid barnatý, anhydrous ≥84%, technická kvalita
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 84% min
CAS No.: 1304-29-6
Molecular Formula: BaO2
Formula Weight: 169.34
Physical Form: Powder
Density: 4.96
Appearance: Tan
Melting Point: 450° dec.
Solubility: Insoluble in water
Merck Reference: 14,989
UN No.: 1449
MDL No.: MFCD00003454
Vzorec:
BaO₂ Hustota: 4,96 Storage Temperature: Ambient |
MDL Number:
MFCD00003454 CAS číslo: 1304-29-6 EINECS: 215-128-4 UN: 1449 ADR: 5.1,II Merck Index: 14,989 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...