Tisk…

Dusičnan barnatý ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Varování

10648.06 10648.14 10648.22 10648.36 10648.88
10648.06EA 1140 CZK
10648.06 10648.14 10648.22 10648.36 10648.88
Dusičnan barnatý ≥99.999% (metals basis), Puratronic®
Dusičnan barnatý

Purity: 99.999% CAS No.: 10022-31-8 Molecular Formula: BaN2O6 Formula Weight: 261.35 Physical Form: Crystalline Density: 3.24 Appearance: White Melting Point: 592° dec. Merck Reference: 14,983 UN No.: 1446 MDL No.: MFCD00003442

Vzorec: Ba(NO₃)₂
Hustota: 3,24
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS číslo: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1,II
Merck Index: 14,983

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR