Tisk…

Lithium titanate

Dodavatel: Thermo Scientific

Synonyms: Lithium titanium oxide, Lithium titanate, Lithium titanate (IV)

13411.18 13411.30
13411.18EA 3190 CZK
13411.18 13411.30
Lithium titanate
Lithium titanate

CAS No.: 12031-82-2 Molecular Formula: Li2TiO3 Formula Weight: 109.78 Physical Form: Powder Odour: Odourless Melting Point: 1520-1564° MDL No.: MFCD00016181

Vzorec: Li₂O₃Ti
MW: 109,76 g/mol
Bod tání: 1520…1564 °C
Storage Temperature: Ambient
MDL Number: MFCD00016181
CAS číslo: 12031-82-2
EINECS: 234-759-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR