Tisk…

Orthotelluric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Telluric acid

M-Clarity™ quality level = MQ200

86375-100G 86375-25G
SIAL86375-100GEA 13500 CZK
SIAL86375-100G SIAL86375-25G
Orthotelluric acid, Sigma-Aldrich®
Orthotelluric acid
Vzorec: H₆O₆Te
MW: 229.64
Bod tání: 136
Hustota: ∼3.068 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011609
CAS číslo: 7803-68-1
EINECS: 232-267-6
UN: 3284
ADR: 6.1 III
Merck Index: 12,09272

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR