Tisk…

Sodium selenate, anhydrous ≥99.8% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

12613.18 12613.06 12613.30 12613.A1
12613.18EA 2260 CZK
12613.18 12613.06 12613.30 12613.A1
Sodium selenate, anhydrous ≥99.8% (metals basis)
Sodium selenate

Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.


Stable under alkaline and oxidizing conditions.


As a reagent

Vzorec: Na₂O₄Se
Hustota: 3,213
Storage Temperature: Ambient
MDL Number: MFCD00003490
CAS číslo: 13410-01-0
EINECS: 236-501-8
UN: 2630
ADR: 6.1,I
Merck Index: 14,8671

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR