Selenious acid ≥99.999% (metals basis), Puratronic®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Selenous acid
10855.14
10855.22
10855.14EA
12560
CZK
10855.14
10855.22
Selenious acid ≥99.999% (metals basis), Puratronic®
Selenious acid
Purity: 99.999%
CAS No.: 7783-00-8
Molecular Formula: H2SeO3
Formula Weight: 128.97
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Merck Reference: 14,8430
UN No.: 3290
MDL No.: MFCD00011331
Vzorec:
H₂O₃Se MW: 128.97 Bod varu: 315 Bod tání: 70 °C (rozkl.) Hustota: 1.998 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011331 CAS číslo: 7783-00-8 EINECS: 231-974-7 UN: 3290 ADR: 6.1 II Merck Index: 14,8430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...