Tisk…

Síran lithný hydrate ≥99.996% (metals basis), Puratronic®

Dodavatel: Thermo Scientific
Varování

12983.06 12983.14
12983.06EA 1820 CZK
12983.06 12983.14
Síran lithný hydrate ≥99.996% (metals basis), Puratronic®
Síran lithný hydrate

Purity: 99.996% CAS No.: 10377-48-7 Molecular Formula: Li2O4S.xH2O Formula Weight: 109.94(anhy) Physical Form: Crystalline Density: 2.06 Appearance: White Melting Point: 130° -H2O Merck Reference: 14,5541 MDL No.: MFCD00011086

Vzorec: Li₂O₄S·xH₂O
MW: 109,9 g/mol (anhy)
Bod varu: 1377 °C (1013 hPa)
Bod tání: 130 °C (-H₂O)
Hustota: 2,06
Storage Temperature: Ambient
MDL Number: MFCD00011086
CAS číslo: 10377-48-7
EINECS: 233-820-4
Merck Index: 14,5541

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR