Tisk…

Sulfit sodný, anhydrous 98-100.0% (by iodometry), Puriss. p.a. Reag. Ph. Eur., Fluka™

Dodavatel: Honeywell Chemicals

31454-1KG 31454-500G 31454-50KG 31454-5KG
31454-1KGEA 1770 CZK
31454-1KG 31454-500G 31454-50KG 31454-5KG
Sulfit sodný, anhydrous 98-100.0% (by iodometry), Puriss. p.a. Reag. Ph. Eur., Fluka™
Sulfit sodný
Vzorec: Na₂SO₃
Bod tání: 600 °C
Hustota: 2,6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS číslo: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Objednat

Výsledky specifikačního testu

Assay (Iodometric) 98 - 100.0 %
Arsenic (As) Max. 0.00002 %
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Selenium (Se) Max. 0.001 %
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.005 %
Thiosulfate (S₂O₃) Max. 0.02 %
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR