thiokyanatan sodný 98,0 ACS
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Sodium rhodanide
33388.36
33388.A3
33388.22
33388.36EA
3220
CZK
33388.36
33388.A3
33388.22
thiokyanatan sodný 98,0 ACS
thiokyanatan sodný
Purity: 98.0% min
CAS No.: 540-72-7
Molecular Formula: NaSCN
Formula Weight: 81.07
Physical Form: Crystalline/Chunks
Density: 1.74
Appearance: White
Odour: Odourless
pH: 6-8
Melting Point: 287°
Solubility: Soluble in water, acetone, alcohol
Beilstein Reference: 3594965
Merck Reference: 14,9327
MDL No.: MFCD00011123
Vzorec:
NaSCN MW: 81.07 Bod tání: 287 Hustota: 1.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011123 CAS číslo: 540-72-7 EINECS: 208-754-4 Merck Index: 14,9327 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...