Tisk…

Disodium hexahydroxoplatinate ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

12620.03 12620.06
12620.03EA 8820 CZK
12620.03 12620.06
Disodium hexahydroxoplatinate ≥99.9% (metals basis)
Disodium hexahydroxoplatinate

Purity: 99.9% CAS No.: 12325-31-4 Molecular Formula: H6Na2O6Pt Formula Weight: 343.10 Physical Form: Crystalline Appearance: Yellow to brown Melting Point: 150° dec. MDL No.: MFCD00014236

Vzorec: H₆Na₂O₆Pt
Storage Temperature: Ambient
MDL Number: MFCD00014236
CAS číslo: 12325-31-4
EINECS: 235-590-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR