Tisk…
Hydrogenfosforečnan dvojsodný, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
M-Clarity™ quality level = MQ300
71642-1KG 71642-5KG
SIAL71642-1KGEA 3170 CZK
SIAL71642-1KG SIAL71642-5KG
Hydrogenfosforečnan dvojsodný, Sigma-Aldrich®
Hydrogenfosforečnan dvojsodný
Vzorec: Na₂HPO₄
MW: 141.96
Bod tání: 250
Hustota: 1.53
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS číslo: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805