Peroxid sodný ≥93% ACS
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
di-Sodium peroxide
Purity: 93% min
CAS No.: 1313-60-6
Molecular Formula: Na2O2
Formula Weight: 77.98
Physical Form: Granular, Packaged in screwtop can
Density: 2.805
Appearance: White to pale yellow
Melting Point: 460° dec.
Solubility: Soluble in water, forming NaOH and H2O2
Merck Reference: 14,8655
UN No.: 1504
MDL No.: MFCD00003497
As an oxidizing agent, a bleaching agent, deOdourant, pharmaceuticals and dyes, and as an analytical reagent
Vzorec:
Na₂O₂ MW: 77.98 Bod tání: 460 °C (rozkl.) Hustota: 2.805 Storage Temperature: Ambient |
MDL Number:
MFCD00003497 CAS číslo: 1313-60-6 EINECS: 215-209-4 UN: 1504 ADR: 5.1 I Merck Index: 14,8655 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...