Tisk…

Voda for molecular biology, sterile filtered, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

95284-100ML 95284-1L 95284-6X100ML
SIAL95284-100MLEA 1620 CZK
SIAL95284-100ML SIAL95284-1L SIAL95284-6X100ML
Voda for molecular biology, sterile filtered, Sigma-Aldrich®
Voda
Vzorec: H₂O
Bod varu: 100 °C (1013 hPa)
Bod tání: 0 °C
Hustota: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS číslo: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR