Tisk…

Dusičnan sodný ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10873.14 10873.22
10873.14EA 7690 CZK
10873.14 10873.22
Dusičnan sodný ≥99.999% (metals basis), Puratronic®
Dusičnan sodný

Purity: 99.999% CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Physical Form: Powder Density: 2.26 Melting Point: 308° Merck Reference: 14,8647 UN No.: 1498 MDL No.: MFCD00011119

Vzorec: NaNO₃
Bod varu: 380 °C (1013 hPa)
Bod tání: 308 °C
Hustota: 2,26
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS číslo: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 14,8647

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR