Tisk…

Dusitan draselný ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L13289.22 L13289.36
L13289.22EA 882 CZK
L13289.22 L13289.36
Dusitan draselný ≥97%
Dusitan draselný

Purity: 97% CAS No.: 7758-09-0 Molecular Formula: KNO2 Formula Weight: 85.11 Odour: Odourless pH: 7.0-10.0 Melting Point: 350° dec. Merck Reference: 14,7649 UN No.: 1488 MDL No.: MFCD00011408

Vzorec: KNO₂
Hustota: 1,915
Storage Temperature: Ambient
MDL Number: MFCD00011408
CAS číslo: 7758-09-0
EINECS: 231-832-4
UN: 1488
ADR: 5.1,II
Merck Index: 14,7649

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR