Lithium amide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
213217-5G
213217-100G
213217-500G
SIAL213217-5GEA
1240
CZK
SIAL213217-5G
SIAL213217-100G
SIAL213217-500G
Lithium amide, Sigma-Aldrich®
Lithium amide
Vzorec:
H₂LiN MW: 22,96 g/mol Bod varu: 430 °C (1013 hPa) Bod tání: 373 °C Hustota: 1,17 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011093 CAS číslo: 7782-89-0 EINECS: 231-968-4 UN: 1390 ADR: 4.3,II Merck Index: 13,05544 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...