Trilithium nitride ≥99.4% (metals basis)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
35479.09
35479.18
35479.09EA
3150
CZK
35479.09
35479.18
Trilithium nitride ≥99.4% (metals basis)
Trilithium nitride
Purity: 99.4%
CAS No.: 26134-62-3
Molecular Formula: Li3N
Formula Weight: 34.82
Physical Form: -60 Mesh Powder
Density: 1.30
Appearance: Red to brown
Odour: Ammonia-like
Melting Point: 845°
UN No.: 2806
MDL No.: MFCD00016186
Vzorec:
Li₃N MW: 34,83 g/mol Bod tání: 845 °C Hustota: 1,30 Storage Temperature: Ambient |
MDL Number:
MFCD00016186 CAS číslo: 26134-62-3 EINECS: 247-475-2 UN: 2806 ADR: 4.3,I |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...