Sodium dicyanoamide, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
178322-25G
178322-100G
SIAL178322-25GEA
1500
CZK
SIAL178322-25G
SIAL178322-100G
Sodium dicyanoamide, Sigma-Aldrich®
Sodium dicyanoamide
Vzorec:
C₂N₃Na Storage Temperature: Ambient |
MDL Number:
MFCD00003535 CAS číslo: 1934-75-4 EINECS: 217-703-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...