Manganistan draselný ≥99.0% ACS low in mercury
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Permanganic acid potassium salt
36675.22
36675.36
36675.A4
36675.22EA
1490
CZK
36675.22
36675.36
36675.A4
Manganistan draselný ≥99.0% ACS low in mercury
Manganistan draselný
Purity: 99.0% min
CAS No.: 7722-64-7
Molecular Formula: KMnO4
Formula Weight: 158.03
Physical Form: Crystalline
Density: 2.7
Appearance: Dark purple
Melting Point: 240° dec.
Merck Reference: 14,7655
UN No.: 1490
MDL No.: MFCD00011364
Vzorec:
KMnO₄ MW: 158,03 g/mol Bod tání: 240 °C (rozkl.) Hustota: 2,7 Storage Temperature: Ambient |
MDL Number:
MFCD00011364 CAS číslo: 7722-64-7 EINECS: 231-760-3 UN: 1490 ADR: 5.1,II Merck Index: 14,7655 |