Tisk…

Disodium hexabromoiridate (≥24,1% Ir)

Dodavatel: Thermo Fisher Scientific
Varování

12649.03 12649.06
12649.03EA 6760 CZK
12649.03 12649.06
Disodium hexabromoiridate (≥24,1% Ir)
Disodium hexabromoiridate

CAS No.: 28529-99-9 Molecular Formula: Na2IrBr6 Formula Weight: 717.63 Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: 350° MDL No.: MFCD00064675

Vzorec: Br₆IrNa₂
Bod tání: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD00064675
CAS číslo: 28529-99-9
EINECS: 249-069-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR