Tisk…

Jodid sodný, ultra dry ≥99.99% (metals basis)

Dodavatel: Thermo Scientific
Varování

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

Ampuled under argon

44766.14 44766.22 44766.36
44766.14EA 5010 CZK
44766.14 44766.22 44766.36
Jodid sodný, ultra dry ≥99.99% (metals basis)
Jodid sodný

Purity: 99.99% CAS No.: 7681-82-5 Molecular Formula: NaI Formula Weight: 149.89 Physical Form: Powder, Ampouled under argon Density: 3.67 Refractive Index: 1.7745 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 651° Boiling Point: 1300-1304° Merck Reference: 14,8631 MDL No.: MFCD00003532

Vzorec: NaI
MW: 149,89 g/mol
Bod varu: 1300…1304 °C
Bod tání: 651 °C
Hustota: 3,67
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS číslo: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 14,8631

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR