Tisk…

Jodid sodný, ultra dry ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování


Ampuled under argon

13604.06 13604.14 13604.22
13604.06EA 2880 CZK
13604.06 13604.14 13604.22
Jodid sodný, ultra dry ≥99.999% (metals basis)
Jodid sodný

Purity: 99.999% CAS No.: 7681-82-5 Molecular Formula: NaI Formula Weight: 149.89 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.67 Refractive Index: 1.7745 Sensitivity: Light Sensitive and Hygroscopic Appearance: White Odour: Odourless Melting Point: 651° Boiling Point: 1300-1304° Merck Reference: 14,8631 MDL No.: MFCD00003532

Vzorec: NaI
Bod varu: 1300…1304 °C
Bod tání: 651 °C
Hustota: 3,67
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS číslo: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 14,8631

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR