Fluorid draselný ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
10980.04
10980.09
10980.18
10980.04EA
1750
CZK
10980.04
10980.09
10980.18
Fluorid draselný ≥99.99% (metals basis)
Fluorid draselný
Purity: 99.99%
CAS No.: 7789-23-3
Molecular Formula: FK
Formula Weight: 58.10
Physical Form: Crystalline
Density: 2.48
Refractive Index: 1.363
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 859°
Boiling Point: 1505°
Beilstein Reference: 3902818
Merck Reference: 14,7632
UN No.: 1812
MDL No.: MFCD00011398
Vzorec:
KF MW: 58.1 Bod varu: 1505 °C Bod tání: 859 °C Hustota: 2.48 Storage Temperature: Ambient |
MDL Number:
MFCD00011398 CAS číslo: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1 III Merck Index: 14,7632 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...