Uhličitan draselný, anhydrous 99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Potash
A16625.0C
A16625.0E
A16625.36
A16625.0CEA
7550
CZK
A16625.0C
A16625.0E
A16625.36
Uhličitan draselný, anhydrous 99%
Uhličitan draselný
Purity: 99%
CAS No.: 584-08-7
Molecular Formula: CK2O3
Formula Weight: 138.21
Odour: Odourless
pH: 11,5-12,5
Melting Point: 891°
Beilstein Reference: 4267587
Merck Reference: 14,7619
MDL No.: MFCD00011382
Vzorec:
K₂CO₃ MW: 138.21 Bod tání: 891 Hustota: 2.42 Storage Temperature: Ambient |
MDL Number:
MFCD00011382 CAS číslo: 584-08-7 EINECS: 209-529-3 Merck Index: 14,7619 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...