Uhličitan draselný ≥99.0% ACS
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Potash
12609.22
12609.36
12609.A1
12609.A7
12609.14
12609.22EA
1340
CZK
12609.22
12609.36
12609.A1
12609.A7
12609.14
Uhličitan draselný ≥99.0% ACS
Uhličitan draselný
Purity: 99.0% min
CAS No.: 584-08-7
Molecular Formula: K2CO3
Formula Weight: 138.21
Physical Form: Granular
Density: 2.29
Melting Point: 891°
Beilstein Reference: 4267587
Merck Reference: 14,7619
MDL No.: MFCD00011382
Vzorec:
K₂CO₃ MW: 138.21 Bod tání: 891 Hustota: 2.42 Storage Temperature: Ambient |
MDL Number:
MFCD00011382 CAS číslo: 584-08-7 EINECS: 209-529-3 Merck Index: 14,7619 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...