Tisk…

broman draselný ≥99.8% ACS

Dodavatel: Thermo Fisher Scientific
Nebezpečí

40013.22 40013.36
40013.22EA 1730 CZK
40013.22 40013.36
broman draselný ≥99.8% ACS
broman draselný

Purity: 99.8% min CAS No.: 7758-01-2 Molecular Formula: KBrO3 Formula Weight: 167.00 Physical Form: Granular Density: 3.27 Melting Point: 370° dec. Solubility: Slightly soluble in cold water. Soluble in boiling water Merck Reference: 14,7617 UN No.: 1484 MDL No.: MFCD00011359


As an analytical reagent, oxidizing agent

Vzorec: KBrO₃
Hustota: 3,27
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS číslo: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II
Merck Index: 14,7617

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR