Tisk…

Tetrafluorboritan sodný 97

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Sodium fluoroborate

A12393.22 A12393.36 A12393.A3
A12393.22EA 858 CZK
A12393.22 A12393.36 A12393.A3
Tetrafluorboritan sodný 97
Tetrafluorboritan sodný

Purity: 97% CAS No.: 13755-29-8 Molecular Formula: BF4Na Formula Weight: 109.79 Melting Point: 384° dec. Merck Reference: 14,8617 MDL No.: MFCD00003515

Vzorec: BF₄Na
MW: 109.79
Bod tání: 384 °C (rozkl.)
Hustota: 2.47
Storage Temperature: Ambient
MDL Number: MFCD00003515
CAS číslo: 13755-29-8
EINECS: 237-340-6
Merck Index: 12,08761

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR