Tisk…

Lithium hexafluoroarsenate ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

11530.09 11530.18
11530.09EA 4910 CZK
11530.09 11530.18
Lithium hexafluoroarsenate ≥99%
Lithium hexafluoroarsenate

Purity: 99% CAS No.: 29935-35-1 Molecular Formula: LiAsF6 Formula Weight: 195.85 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 1557 MDL No.: MFCD00011076

Vzorec: AsF₆Li
Bod tání: 349 °C
Storage Temperature: Ambient
MDL Number: MFCD00011076
CAS číslo: 29935-35-1
EINECS: 249-963-0
UN: 1557
ADR: 6.1,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR