Methylcyclopentane, Supelco®
Dodavatel: Merck
MQ100
66490-10ML
66490-50ML
66490-10MLEA
7840
CZK
66490-10ML
66490-50ML
Methylcyclopentane, Supelco®
Methylcyclopentane
Vzorec:
C₆H₁₂ MW: 84.16 Bod varu: 71.8 Bod tání: –142 °C Hustota: 0.74 g/cm³ (20 °C) Bod vzplanutí: –17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001382 CAS číslo: 96-37-7 EINECS: 202-503-2 UN: 2298 ADR: 3 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...