trans-4-Methylcyclohexanol ≥98%
Dodavatel: Thermo Fisher Scientific
B20054.06
B20054.14
B20054.06EA
3360
CZK
B20054.06
B20054.14
trans-4-Methylcyclohexanol ≥98%
trans-4-Methylcyclohexanol
Purity: 98%
CAS No.: 7731-29-5
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 72°(162°F)
Refractive Index: 1.4550
Boiling Point: 173-175°
MDL No.: MFCD00064171
Vzorec:
C₇H₁₄O |
MDL Number:
MFCD00064171 CAS číslo: 7731-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...