Cyclopropanemethylamine 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Cyclopropylmethylamine, (Cyclopropylmethyl)amine, (Aminomethyl)cyclopropane
258720100
258720050
258720500
ACRO258720100EA
5450
CZK
ACRO258720100
ACRO258720050
ACRO258720500
Cyclopropanemethylamine 97%
Cyclopropanemethylamine
Vzorec:
C₄H₉N MW: 71,12 g/mol Bod varu: 84…85 °C (760 mmHg) Hustota: 0,82 g/cm³ Bod vzplanutí: –30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037147 CAS číslo: 2516-47-4 EINECS: 219-737-6 UN: 2733 ADR: 3,II |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4300 to 1.4340 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...