Dimedon ≥98%
Dodavatel: Thermo Scientific
Synonyms:
5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethylcyclohexane-1,3-dione
A10140.14
A10140.22
A10140.36
A10140.14EA
978
CZK
A10140.14
A10140.22
A10140.36
Dimedon ≥98%
Dimedon
Purity: 98%
CAS No.: 126-81-8
Molecular Formula: C8H12O2
Formula Weight: 140.18
Melting Point: 146-150°
Beilstein Reference: 471489
Merck Reference: 14,3242
MDL No.: MFCD00001588
Vzorec:
C₈H₁₂O₂ MW: 140,18 g/mol Bod varu: 234 °C (1013 hPa) Bod tání: 146…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001588 CAS číslo: 126-81-8 EINECS: 204-804-4 Merck Index: 14,3242 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...