Tisk…

2-Ethyl-1,3-cyclopentanedione ≥99%

Dodavatel: Thermo Fisher Scientific

B20254.06 B20254.14
B20254.06EA 1620 CZK
B20254.06 B20254.14
2-Ethyl-1,3-cyclopentanedione ≥99%
2-Ethyl-1,3-cyclopentanedione

Purity: 99% CAS No.: 823-36-9 Molecular Formula: C7H10O2 Formula Weight: 126.16 Storage Temperature: -30°C to -10°C Melting Point: 168-170° Beilstein Reference: 1860068 MDL No.: MFCD00044191

Vzorec: C₇H₁₀O₂
Bod tání: 168…170 °C
MDL Number: MFCD00044191
CAS číslo: 823-36-9
EINECS: 212-512-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR