Tisk…

4-tert-Pentylcyclohexanone ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-(tert-Pentyl)cyclohexanone, 4-tert-Amylcyclohexanone

A12832.18 A12832.30
A12832.18EA 2210 CZK
A12832.18 A12832.30
4-tert-Pentylcyclohexanone ≥97%
4-tert-Pentylcyclohexanone

Purity: 97% CAS No.: 16587-71-6 Molecular Formula: C11H20O Formula Weight: 168.28 Refractive Index: 1.4690 Boiling Point: 124-125°/16mm MDL No.: MFCD00013798

Vzorec: C₁₁H₂₀O
MW: 168.28
Bod varu: 124…125 °C (16 mmHg)
Bod tání: 96
Hustota: 0.92
Storage Temperature: Ambient
MDL Number: MFCD00013798
CAS číslo: 16587-71-6
EINECS: 240-642-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR