2,2,6-Trimethylcyclohexanone ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
B21111.03
B21111.06
B21111.14
B21111.03EA
2310
CZK
B21111.03
B21111.06
B21111.14
2,2,6-Trimethylcyclohexanone ≥97%
2,2,6-Trimethylcyclohexanone
Purity: 97%
CAS No.: 2408-37-9
Molecular Formula: C9H16O
Formula Weight: 140.23
Flash Point: 51°(125°F)
Refractive Index: 1.4470
Boiling Point: 178-179°
Beilstein Reference: 1905593
UN No.: 1224
MDL No.: MFCD00045550
Vzorec:
C₉H₁₆O MW: 140,23 g/mol Bod varu: 178…179 °C Bod tání: –31,8 °C Hustota: 0,904 Bod vzplanutí: 51 °C (125 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00045550 CAS číslo: 2408-37-9 EINECS: 219-309-9 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...