Ethyl-2-furoate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Ethyl2-furoate, Ethyl furan-2-carboxylate
M-Clarity™ quality level = MQ300
W501301-100G-K
W501301-1KG-K
SIALW501301-100G-KEA
0
CZK
SIALW501301-100G-K
SIALW501301-1KG-K
Ethyl-2-furoate, Sigma-Aldrich®
Ethyl-2-furoate
Vzorec:
C₇H₈O₃ MW: 140,14 g/mol |
MDL Number:
MFCD00003237 CAS číslo: 614-99-3 EINECS: 210-404-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...