Tisk…

(±)-α-Terpineol, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

M-Clarity™ quality level = MQ100

W304522-10KG-K W304522-1KG-K W304522-5KG-K
SIALW304522-10KG-KEA 24600 CZK
SIALW304522-10KG-K SIALW304522-1KG-K SIALW304522-5KG-K
(±)-α-Terpineol, Sigma-Aldrich®
(±)-α-Terpineol
Vzorec: C₁₀H₁₈O
MW: 154.25
MDL Number: MFCD00001557
CAS číslo: 98-55-5
EINECS: 202-680-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR