Tisk…

TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid

T9659-100G T9659-1KG T9659-250G
SIALT9659-100GEA 10120 CZK
SIALT9659-100G SIALT9659-1KG SIALT9659-250G
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Sigma-Aldrich®
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Vzorec: C₇H₁₇NO₆S
MW: 243,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007538
CAS číslo: 29915-38-6
EINECS: 249-954-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR