TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid
T5316-100G
T5316-1KG
T5316-25G
T5316-500G
SIALT5316-100GEA
0
CZK
SIALT5316-100G
SIALT5316-1KG
SIALT5316-25G
SIALT5316-500G
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Sigma-Aldrich®
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Vzorec:
C₇H₁₇NO₆S MW: 243,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007538 CAS číslo: 29915-38-6 EINECS: 249-954-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...