Tisk…

Saccharin sodium hydrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

S6047-1KG S6047-250G
SIALS6047-1KGEA 12550 CZK
SIALS6047-1KG SIALS6047-250G
Saccharin sodium hydrate, Sigma-Aldrich®
Saccharin sodium hydrate
Vzorec: C₇H₅NNaO₃S·xH₂O
MW: 205,17 g/mol (anhy)
MDL Number: MFCD00149605
CAS číslo: 82385-42-0
EINECS: 204-886-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR