Saccharin sodium hydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
S1002-1KG
S1002-500G
SIALS1002-1KGEA
5540
CZK
SIALS1002-1KG
SIALS1002-500G
Saccharin sodium hydrate, Sigma-Aldrich®
Saccharin sodium hydrate
Vzorec:
C₇H₅NNaO₃S·xH₂O MW: 205,17 g/mol (anhy) |
MDL Number:
MFCD00149605 CAS číslo: 82385-42-0 EINECS: 204-886-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...