Tisk…

Brij® 96, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

P6136-100G P6136-1KG P6136-500G
SIALP6136-100GEA 1670 CZK
SIALP6136-100G SIALP6136-1KG SIALP6136-500G
Brij® 96, Sigma-Aldrich®
Brij® 96
Vzorec: C₃₈H₇₆O₁₁
CAS číslo: 9004-98-2
EINECS: 500-016-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR