Tisk…

Brij® 58, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Polyoxyethylene (20) cetyl ether, Polyethylene glycol hexadecyl ether

M-Clarity™ quality level = MQ200

P5884-100G P5884-1KG P5884-500G
SIALP5884-100GEA 2220 CZK
SIALP5884-100G SIALP5884-1KG SIALP5884-500G
Brij® 58, Sigma-Aldrich®
Brij® 58
Vzorec: C₅₆H₁₁₄O₂₁
MW: 1123,51 g/mol
MDL Number: MFCD00080892
CAS číslo: 9004-95-9
EINECS: 500-014-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR