Tisk…

2-Furoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Furan-2-carboxylic acid, 2-Furancarboxylic acid

M-Clarity™ quality level = MQ100

F20505-100G F20505-500G F20505-5G
SIALF20505-100GEA 1190 CZK
SIALF20505-100G SIALF20505-500G SIALF20505-5G
2-Furoic acid, Sigma-Aldrich®
2-Furoic acid
Vzorec: C₅H₄O₃
MW: 112.08
MDL Number: MFCD00003238
CAS číslo: 88-14-2
EINECS: 201-803-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR