Tisk…

4-Chloropyridine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Chloropyridinium chloride

M-Clarity™ quality level = MQ100

C70223-100G C70223-25G
SIALC70223-100GEA 7970 CZK
SIALC70223-100G SIALC70223-25G
4-Chloropyridine hydrochloride, Sigma-Aldrich®
4-Chloropyridine hydrochloride
Vzorec: C₅H₅Cl₂N
MDL Number: MFCD00012829
CAS číslo: 7379-35-3
EINECS: 230-946-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR